Learn More
Invitrogen™ IHH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593107
Description
Immunogen sequence: GCFPAGAQVR LESGARVALS AVRPGDRVLA MGEDGSPTFS DVLIFLDREP HRLRAFQVIE TQDPPRRLAL TPAHLLFTAD NHTEPAARFR ATFASHVQPG QYVLVAGVPG LQPARVAAVS THVALGAYAP LTKHGTLVVE DVVASCFAAV ADHHLAQLAF WPLRLFHSLA WGSWTPGEGV HWYPQLLYRL GRLLLEEGSF HPLGMSGAGS; Positive Samples: Human placenta, Mouse kidney, Mouse liver, Mouse ovary, Rat liver, Rat lung, Rat brain; Cellular Location: Cell membrane, Extracellular side, Lipid-anchor, extracellular space.
Ihh encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.
Specifications
IHH | |
Polyclonal | |
Unconjugated | |
Ihh | |
BDA1; HHG2; HHG-2; Ihh; Indian hedgehog; Indian hedgehog homolog; indian hedgehog protein; Indian hedgehog protein C-product; Indian hedgehog protein N-product; indian hedgehog protein; LOW QUALITY PROTEIN: indian hedgehog protein; Indian hedgehog signaling molecule; Indian hedgehog-like protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16147, 3549, 84399 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97812, Q14623 | |
Ihh | |
Recombinant fusion protein containing a sequence corresponding to amino acids 202-411 of human IHH (NP_0021722). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.