Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IIGP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572990
Description
IIGP1 Polyclonal Antibody for Western Blot
GTPase with low activity. Has higher affinity for GDP than for GTP. Plays a role in resistance to intracellular pathogens. Required for disruption of the parasitophorous vacuole formed following T.gondii infection and subsequent killing of the parasite. Mediates resistance to C.trachomatis infection by targeting bacterial inclusions to autophagosomes for subsequent lysosomal destruction.
Specifications
IIGP1 | |
Polyclonal | |
Unconjugated | |
Iigp1 | |
2900074L10Rik; AI046432; AW111922; Ifgga1; Iigp; Iigp1; interferon inducible GTPase 1; interferon-gamma-inducible GTPase Ifgga1 protein; interferon-inducible GTPase 1; Irga6 | |
Rabbit | |
Protein A | |
RUO | |
60440 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q9QZ85 | |
Iigp1 | |
A portion of amino acids 325-375 of mouse Iigp1 was used as the immunogen for this antibody. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction