Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IKAP Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50 - $596.50
Specifications
Antigen | IKAP |
---|---|
Concentration | 0.05 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111296
|
Invitrogen™
PA5111296 |
100 μL |
Each for $596.50
|
|
|||||
PIPA5111320
|
Invitrogen™
PA5111320 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: NLKDEVYHIL KVLFLFEFDE QGRELQKAFE DTLQLMERSL PEIWTLTYQQ NSATPVLGPN STANSIMASY QQQKTSVPVL DAELFIPPKI NRRTQWKLSL LD.
The protein encoded by this gene is a scaffold protein and a regulator for 3 different kinases involved in proinflammatory signaling. This encoded protein can bind NF-kappa-B-inducing kinase (NIK) and IKKs through separate domains and assemble them into an active kinase complex. Mutations in this gene have been associated with familial dysautonomia.Specifications
IKAP | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
ELP1 | |
3110040G09Rik; 6030413P05; C78473; DKFZp781H1425; DYS; elongator acetyltransferase complex subunit 1; elongator complex protein 1; Elp1; FD; FLJ12497; IKAP; IkappaB kinase complex-associated protein; IKBKAP; IKI3; IKK complex-associated protein; IKK-complex-associated protein IKAP; inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein; inhibitor of kappa light polypeptide enhancer in B-cells, kinase complex-associated protein; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein; p150; RP11-3J11.4; TOT1 | |
ELP1 | |
Primary | |
Antigen Affinity Chromatography |
0.05 mg/mL | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95163 | |
8518 | |
Recombinant Protein corresonding to Human IKAP. Recombinant protein control fragment (Product #RP-104882). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title