Learn More
Invitrogen™ IKAP Polyclonal Antibody

Description
PA1-4164 detects IKAP in human samples. PA1-4164 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~158 kDa protein representing IKAP in Interleukin-1-stimulated 293 cells. The PA1-4164 immunogen is a synthetic peptide corresponding to the C-terminus of IKAP.
Specifications
Specifications
| Antigen | IKAP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ELP1 |
| Gene Accession No. | O95163 |
| Gene Alias | 3110040G09Rik; 6030413P05; C78473; DKFZp781H1425; DYS; elongator acetyltransferase complex subunit 1; elongator complex protein 1; Elp1; FD; FLJ12497; IKAP; IkappaB kinase complex-associated protein; IKBKAP; IKI3; IKK complex-associated protein; IKK-complex-associated protein IKAP; inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein; inhibitor of kappa light polypeptide enhancer in B-cells, kinase complex-associated protein; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein; p150; RP11-3J11.4; TOT1 |
| Gene Symbols | ELP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.