Learn More
Invitrogen™ IKAP Polyclonal Antibody

Description
Immunogen sequence: RLHVLCQGWH YLAYDWHWTT DRSVGDNSSD LSNVAVIDGN RVLVTVFRQT VVPPPMCTYQ LLFPHPVNQV TFLAHPQKSN DLAVLDASNQ ISVYKCGDCP SADPTVKLGA VG Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 77%.
Specifications
Specifications
| Antigen | IKAP |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ELP1 |
| Gene Accession No. | O95163 |
| Gene Alias | 3110040G09Rik; 6030413P05; C78473; DKFZp781H1425; DYS; elongator acetyltransferase complex subunit 1; elongator complex protein 1; Elp1; FD; FLJ12497; IKAP; IkappaB kinase complex-associated protein; IKBKAP; IKI3; IKK complex-associated protein; IKK-complex-associated protein IKAP; inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein; inhibitor of kappa light polypeptide enhancer in B-cells, kinase complex-associated protein; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein; p150; RP11-3J11.4; TOT1 |
| Gene Symbols | ELP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.