Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IKAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144315
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144315 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144315 Supplier Invitrogen™ Supplier No. PA5144315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IKAP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA

The protein encoded by this gene is a scaffold protein and a regulator for 3 different kinases involved in proinflammatory signaling. This encoded protein can bind NF-kappa-B-inducing kinase (NIK) and IKKs through separate domains and assemble them into an active kinase complex. Mutations in this gene have been associated with familial dysautonomia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKAP
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.665 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ELP1
Gene Accession No. O95163
Gene Alias 3110040G09Rik; 6030413P05; C78473; DKFZp781H1425; DYS; elongator acetyltransferase complex subunit 1; elongator complex protein 1; Elp1; FD; FLJ12497; IKAP; IkappaB kinase complex-associated protein; IKBKAP; IKI3; IKK complex-associated protein; IKK-complex-associated protein IKAP; inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein; inhibitor of kappa light polypeptide enhancer in B-cells, kinase complex-associated protein; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein; p150; RP11-3J11.4; TOT1
Gene Symbols ELP1
Host Species Rabbit
Immunogen Recombinant Human Elongator complex protein 1 protein (1107-1258AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8518
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.