Learn More
IKAP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303163AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a scaffold protein and a regulator for 3 different kinases involved in proinflammatory signaling. This encoded protein can bind NF-kappa-B-inducing kinase (NIK) and IKKs through separate domains and assemble them into an active kinase complex. Mutations in this gene have been associated with familial dysautonomia.Specifications
IKAP | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95163 | |
ELP1 | |
Between 1150 and 1200 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ELP1 | |
IkappaB kinase complex-associated protein, IKK complex-associated protein, p150 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8518 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.