Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ikaros/IKZF1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP100125
Description
Ikaros/IKZF1 Polyclonal specifically detects Ikaros/IKZF1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
Ikaros/IKZF1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DNA-binding protein Ikaros, hIk-1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, LYF1PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1CLL-associated antigen KW-6 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize all reported human isoforms. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_006051.1 | |
IKZF1 | |
Peptide with sequence C-DLLRAASENSQDALR corresponding to internal region according to NP_006051.1, NP_001207694.1, NP_001207695.1, NP_001207696.1, NP_001207697.1, NP_001207698.1, NP_001207699.1, NP_001207700.1. | |
0.1 mg | |
Innate Immunity | |
10320 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction