Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IKAROS Monoclonal Antibody (4E9), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528612
Description
This antibody recognizes Ikaros, a transcription factor (intracellular antigen) expressed broadly in hematopoietic progenitors and serving as a key regulator of lymphopoiesis. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
DNA-binding protein IKAROS functions in the specification and the maturation of the T lymphocyte. It also interacts with a critical control element in the terminal deoxynucleotidyl transferase promoter as well as with the promoters for other genes expressed during early stages of B-cell and T-cell development.
Specifications
IKAROS | |
Monoclonal | |
PE | |
IKZF1 | |
5832432G11Rik; CLL-associated antigen KW-6; CVID13; DNA-binding protein Ikaros; hIk-1; hlk-1; Hs.54452; IK1; IKAROS; IKAROS family zinc finger 1; IKAROS family zinc finger 1 (Ikaros); ikaros family zinc finger protein 1; IKZF1; KAROS family zinc finger 1 (Ikaros); LYF1; LyF-1; Lymphoid transcription factor LyF-1; mKIAA4227; PPP1R92; PRO0758; protein phosphatase 1, regulatory subunit 92; RGD1562979; Zfpn1a1; zinc finger containing transcription factor; zinc finger protein, subfamily 1A, 1 (Ikaros); Znfn1a1 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
10320, 22778 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
4E9 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q03267, Q13422 | |
IKZF1 | |
Recombinant human Ikaros (C-terminal region). | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction