Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IKAROS Recombinant Rabbit Monoclonal Antibody (4S10I3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535380
Description
Immunogen sequence: AEDLCKIGSE RSLVLDRLAS NVAKRKSSMP QKFLGDKGLS DTPYDSSASY EKENEMMKSH VMDQAINNAI NYLGAESLRP LVQTPPGGSE VVPVISPMYQ L.
DNA-binding protein IKAROS functions in the specification and the maturation of the T lymphocyte. It also interacts with a critical control element in the terminal deoxynucleotidyl transferase promoter as well as with the promoters for other genes expressed during early stages of B-cell and T-cell development.
Specifications
IKAROS | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03267, Q13422 | |
IKZF1 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human Ikaros (Q13422). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ChIP Assay, ELISA, Western Blot, CUT&RUN, Immunocytochemistry | |
4S10I3 | |
Unconjugated | |
IKZF1 | |
5832432G11Rik; CLL-associated antigen KW-6; CVID13; DNA-binding protein Ikaros; hIk-1; hlk-1; Hs.54452; IK1; IKAROS; IKAROS family zinc finger 1; IKAROS family zinc finger 1 (Ikaros); ikaros family zinc finger protein 1; IKZF1; KAROS family zinc finger 1 (Ikaros); LYF1; LyF-1; Lymphoid transcription factor LyF-1; mKIAA4227; PPP1R92; PRO0758; protein phosphatase 1, regulatory subunit 92; RGD1562979; Zfpn1a1; zinc finger containing transcription factor; zinc finger protein, subfamily 1A, 1 (Ikaros); Znfn1a1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10320, 22778 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction