Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IKAROS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519214
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519214 Supplier Invitrogen™ Supplier No. PA519214
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.

DNA-binding protein IKAROS functions in the specification and the maturation of the T lymphocyte. It also interacts with a critical control element in the terminal deoxynucleotidyl transferase promoter as well as with the promoters for other genes expressed during early stages of B-cell and T-cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKAROS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene IKZF1
Gene Accession No. Q13422
Gene Alias 5832432G11Rik; CLL-associated antigen KW-6; CVID13; DNA-binding protein Ikaros; hIk-1; hlk-1; Hs.54452; IK1; IKAROS; IKAROS family zinc finger 1; IKAROS family zinc finger 1 (Ikaros); ikaros family zinc finger protein 1; IKZF1; KAROS family zinc finger 1 (Ikaros); LYF1; LyF-1; Lymphoid transcription factor LyF-1; mKIAA4227; PPP1R92; PRO0758; protein phosphatase 1, regulatory subunit 92; RGD1562979; Zfpn1a1; zinc finger containing transcription factor; zinc finger protein, subfamily 1A, 1 (Ikaros); Znfn1a1
Gene Symbols IKZF1
Host Species Goat
Immunogen Synthetic peptide sequence (DTPYDSSASYEKEN) corresponding to the internal amino acids of IKZF1 (aa 290-304).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10320
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.