Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-alpha Antibody (116), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290043PCP
Description
IkB-alpha Monoclonal antibody specifically detects IkB-alpha in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
IkB-alpha | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
116 | |
Western Blot, ELISA, Immunoprecipitation | |
alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IkB-alpha (Phe2-Leu317; Accession#: NP_065390.1) | |
0.1 mL | |
Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction | |
4792 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction