Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-alpha Antibody (6A920), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056507APC
Description
IkB-alpha Monoclonal specifically detects IkB-alpha in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
IkB-alpha | |
Monoclonal | |
APC | |
P25963 | |
NFKBIA | |
A recombinant protein corresponding to amino acid residues 32-291 of human IkBa was used as immunogen. | |
0.1 mL | |
Apoptosis, Cell Biology, Diabetes Research, DNA Repair, Nucleotide Excision Repair, Signal Transduction | |
4792 | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
6A920 | |
Flow Cytometry | |
alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
A 40 kDa band is observed. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction