Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-alpha Antibody (6A920) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280787
Description
IkB-alpha Monoclonal specifically detects IkB-alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready, Knockout Validated.Specifications
IkB-alpha | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 μg/mL, Simple Western : 1:20, Flow Cytometry 0.25-1 ug/10^6 cells, Immunohistochemistry : 1:20-1:1000, Immunocytochemistry/Immunofluorescence : 1:20-1:1000, Immunoprecipitation : 1 μg/mL, Immunohistochemistry-Paraffin : 1:100, CyTOF-ready, Knockout Validated | |
alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Rat reactivity reported in the scientific literature (PMID: 23840265). | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
6A920 | |
Unconjugated | |
PBS | |
NFKBIA | |
A recombinant protein corresponding to amino acid residues 32-291 of human IkBa was used as immunogen. | |
0.1 mg | |
Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction | |
4792 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction