Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$207.00 - $426.00
Specifications
| Antigen | IkB-alpha |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1 - 3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB12022071T
![]() |
Novus Biologicals
NB12022071SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB12022071
![]() |
Novus Biologicals
NB12022071 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
IkB-alpha Polyclonal specifically detects IkB-alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IkB-alpha | |
| Western Blot 1 - 3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
| NFKBIA | |
| IgG | |
| Protein G purified | |
| In Jurkat, a 40 kDa band is observed. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P25963 | |
| 4792 | |
| A synthetic peptide corresponding to amino acids 34-48 of human IkB-alpha was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title