Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB-alpha, p Ser32, p Ser36 Antibody, Novus Biologicals™
SDP

Catalog No. p-7199818 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017850 0.025 mL
NB10081987 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017850 Supplier Novus Biologicals Supplier No. NB100819870.025ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IkB-alpha Polyclonal specifically detects IkB-alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen IkB-alpha (p Ser32, p Ser36)
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Accession No. P25963
Gene Alias alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
Gene Symbols NFKBIA
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human IkB-alpha. around the phosphorylation site of serine 32/36 (H-D-SP-G-L-D-SP -M-K).
Molecular Weight of Antigen 39 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Diabetes Research, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4792
Test Specificity IkB-alpha. (specific to Phospho-Ser32/Ser36) detects endogenous levels of IkB-alpha only when phosphorylated at serine 32/36.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.