Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-alpha, p Ser32, p Ser36 Antibody (39A1413) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 12 publications
$243.50 - $560.50
Specifications
| Antigen | IkB-alpha (p Ser32, p Ser36) |
|---|---|
| Clone | 39A1413 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, Simple Western 1:50, Immunoprecipitation 1-2 μg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056724T
![]() |
Novus Biologicals
NB10056724SS |
0.025 mg |
Each for $243.50
|
|
|||||
NB10056724
![]() |
Novus Biologicals
NB10056724 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
IkB-alpha Monoclonal specifically detects IkB-alpha in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.Specifications
| IkB-alpha (p Ser32, p Ser36) | |
| 1.0 mg/mL | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Chaperone Mediated Autophagy (CMA), Phospho Specific | |
| P25963 | |
| 4792 | |
| This monoclonal antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 32 and 36 of human IkBa. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 39A1413 | |
| Western Blot 1-2 μg/mL, Simple Western 1:50, Immunoprecipitation 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Canine | |
| alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
| NFKBIA | |
| IgG1 κ | |
| Protein G purified | |
| IkBa (39A1413 ) [Ser32/Ser36] |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title