Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-alpha, p Ser36 Antibody (39A1413), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056724AF405
Description
IkB-alpha Monoclonal specifically detects IkB-alpha in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot.Specifications
IkB-alpha (p Ser32, p Ser36) | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
IkBa (39A1413 ) [Ser32/Ser36] | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot | |
39A1413 | |
Western Blot | |
P25963 | |
NFKBIA | |
This monoclonal antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 32 and 36 of human IkBa. | |
0.1 mL | |
Apoptosis, Cell Biology, Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction | |
4792 | |
Human, Mouse, Pig, Bovine, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction