Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-alpha, p Ser36 Antibody (39A1413), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056724AF532
Description
IkB-alpha Monoclonal specifically detects IkB-alpha in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot.Specifications
IkB-alpha | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
NFKBIA | |
This monoclonal antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 32 and 36 of human IkBa. | |
Primary | |
IkBa (39A1413 ) [Ser32/Ser36] | |
Store at 4°C in the dark. |
Western Blot | |
39A1413 | |
Western Blot | |
alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
Mouse | |
0.1 mL | |
4792 | |
Human, Mouse, Pig | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction