Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB-alpha, p Ser36 Antibody (SR1657), Novus Biologicals™
SDP

Catalog No. NB259129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259129 50 μg
NB258849 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259129 Supplier Novus Biologicals Supplier No. NBP32159450UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IkB-alpha Monoclonal antibody specifically detects IkB-alpha in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen IkB-alpha
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone SR1657
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
Host Species Rabbit
Immunogen A synthesized peptide derived from human phospho-IkB-alpha [Ser36] (Uniprot #: P25963)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4792
Target Species Human, Mouse
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.