Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-alpha, p Tyr42 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323389
Description
IkB-alpha Polyclonal antibody specifically detects IkB-alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
IkB-alpha | |
Polyclonal | |
Western Blot 1:1000, Immunoprecipitation 1:100 | |
alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
IkB-alpha (Tyr-42) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 42 of human IkB-alpha. This peptide sequence has low homology to other I?B proteins, but does have some homology to unrelated proteins that may have a conserved tyrosine phosphorylation motif. [UniProt# P25963] | |
0.1 mL | |
Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction | |
4792 | |
Store at -20°C. | |
IgG |
Western Blot, Immunoprecipitation | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction