Learn More
Invitrogen™ IkB beta Monoclonal Antibody (7B4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515673
Description
MA5-15673 targets NFKBIB in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15673 immunogen is purified recombinant fragment of human NFKBIB expressed in E. Coli. MA5-15673 detects NFKBIB which has a predicted molecular weight of approximately 45kDa.
The NF-kB/Rel transcription factors are present in the cytosol in an inactive state, complexed with the inhibitory IkB proteins. Most agents that activate NF-kB do so through a common pathway based on phosphorylation-induced, proteasome-mediated degradation of IkB. The key regulatory step in this pathway involves activation of a high molecular weight IkB kinase (IKK) complex whose catalysis is generally carried out by three tightly associated IKK subunits. IKK alpha and IKK beta serve as the catalytic subunits of the kinase and IKK gamma serves as the regulatory subunit. Activation of IKK depends upon phosphorylation at Ser177 and Ser181 in the activation loop of IKK beta (Ser176 and Ser180 in IKK alpha), which causes conformational changes, resulting in kinase activation.
Specifications
IkB beta | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q15653 | |
NFKBIB | |
Purified recombinant fragment of human NFKBIB expressed in E. Coli. | |
RUO | |
4793 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
7B4 | |
Unconjugated | |
NFKBIB | |
I kappa B beta; I(Kappa)B(beta); ikappaBbeta; I-kappa-B-beta; IkB; IKB B; IKB beta; Ikbb; ikB-B; IkBbeta; ikB-beta; NF kappa BIB; NF-kappa-B inhibitor beta; NF-kappa-BIB; NFKB inhibitor beta; NFKBIB; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta; Thyroid receptor; thyroid receptor-interacting protein 9; TR-interacting protein 9; TRIP 9; TRIP9; TRIP-9 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.