Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IkB beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA132136
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132136 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132136 Supplier Invitrogen™ Supplier No. PA132136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A control peptide is available, GTX27548.

The NF-kB/Rel transcription factors are present in the cytosol in an inactive state, complexed with the inhibitory IkB proteins. Most agents that activate NF-kB do so through a common pathway based on phosphorylation-induced, proteasome-mediated degradation of IkB. The key regulatory step in this pathway involves activation of a high molecular weight IkB kinase (IKK) complex whose catalysis is generally carried out by three tightly associated IKK subunits. IKK alpha and IKK beta serve as the catalytic subunits of the kinase and IKK gamma serves as the regulatory subunit. Activation of IKK depends upon phosphorylation at Ser177 and Ser181 in the activation loop of IKK beta (Ser176 and Ser180 in IKK alpha), which causes conformational changes, resulting in kinase activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IkB beta
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation Whole serum with 0.01% sodium azide
Gene NFKBIB
Gene Accession No. Q15653, Q60778, Q9JIA3
Gene Alias I kappa B beta; I(Kappa)B(beta); ikappaBbeta; I-kappa-B-beta; IkB; IKB B; IKB beta; Ikbb; ikB-B; IkBbeta; ikB-beta; NF kappa BIB; NF-kappa-B inhibitor beta; NF-kappa-BIB; NFKB inhibitor beta; NFKBIB; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta; Thyroid receptor; thyroid receptor-interacting protein 9; TR-interacting protein 9; TRIP 9; TRIP9; TRIP-9
Gene Symbols NFKBIB
Host Species Rabbit
Immunogen IkBb peptide corresponding to a region near the C-terminus of the human protein conjugated to KLH.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18036, 4793, 81525
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.