Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IkB beta Recombinant Rabbit Monoclonal Antibody (SN0648)
GREENER_CHOICE

Catalog No. PIMA532261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532261 Supplier Invitrogen™ Supplier No. MA532261
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The NF-kB/Rel transcription factors are present in the cytosol in an inactive state, complexed with the inhibitory IkB proteins. Most agents that activate NF-kB do so through a common pathway based on phosphorylation-induced, proteasome-mediated degradation of IkB. The key regulatory step in this pathway involves activation of a high molecular weight IkB kinase (IKK) complex whose catalysis is generally carried out by three tightly associated IKK subunits. IKK alpha and IKK beta serve as the catalytic subunits of the kinase and IKK gamma serves as the regulatory subunit. Activation of IKK depends upon phosphorylation at Ser177 and Ser181 in the activation loop of IKK beta (Ser176 and Ser180 in IKK alpha), which causes conformational changes, resulting in kinase activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IkB beta
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone SN0648
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene NFKBIB
Gene Accession No. Q15653
Gene Alias I kappa B beta; I(Kappa)B(beta); ikappaBbeta; I-kappa-B-beta; IkB; IKB B; IKB beta; Ikbb; ikB-B; IkBbeta; ikB-beta; NF kappa BIB; NF-kappa-B inhibitor beta; NF-kappa-BIB; NFKB inhibitor beta; NFKBIB; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta; Thyroid receptor; thyroid receptor-interacting protein 9; TR-interacting protein 9; TRIP 9; TRIP9; TRIP-9
Gene Symbols NFKBIB
Host Species Rabbit
Immunogen Synthetic peptide within Human IKB beta aa 322-356.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.