Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IkB epsilon Monoclonal Antibody (OTI6H8)
GREENER_CHOICE

Catalog No. PIMA527351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527351 Supplier Invitrogen™ Supplier No. MA527351
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IkB epsilon Monoclonal Antibody for Western Blot, IHC (P)

NFKB1 or NFKB2 is bound to REL, RELA, or RELB to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008 or NFKBIB, MIM 604495), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664, or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine). For some genes, activation requires NFKB interaction with other transcription factors, such as STAT (see STAT6, MIM 601512), AP1 (JUN, MIM 165160), and NFAT (see NFATC1, MIM 600489).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IkB epsilon
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Nfkbie
Gene Accession No. O00221
Gene Alias ikappaBepsilon; I-kappa-B-epsilon; IkB epsilon; IKBE; IkB-E; IkBepsilon; ikB-epsilon; NF-kappa-B inhibitor epsilon; NF-kappa-BIE; NFKB inhibitor epsilon; NFKBIE; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, epsilon; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, epsilon; Slc35b2; solute carrier family 35, member B2
Gene Symbols Nfkbie
Host Species Mouse
Immunogen Full length human recombinant protein of NFKBIE produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4794
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.