Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ IκB Ubiquitination Inhibitor, GS143
Click to view available options
Quantity:
10 mg
Description
- Packaged Under Inert Gas
- Cell Permeable
- Deactivate with 5% Bleach

Specifications
Specifications
Color | Dark Red |
Description | 97% by HPLC |
Target Proteins | phosphorylated IκBα |
Molecular Formula | C28H19FN2O4 |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Powder |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction