Learn More
Invitrogen™ IkB zeta Monoclonal Antibody (LK2NAP), PerCP-eFluor™ 710, eBioscience™
Rat Monoclonal Antibody
$476.00
Specifications
Antigen | IkB zeta |
---|---|
Clone | LK2NAP |
Concentration | 0.2 mg/mL |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
46-680-182
![]() |
Invitrogen™
46680182 |
100 μg |
Each of 1 for $476.00
|
![]() |
|||||
Description
Description: The LK2NAP monoclonal antibody reacts with mouse IkB zeta (INAP, MAIL). IkBz is a member of the IkB family of cytoplasmic proteins and transcription factors and also shares structural homology with Bcl-3. It is induced in monocytes, macrophages and B cells in response to TLR and IL-1R signaling. In NK cells, IkBz is rapidly upregulated in response to IL-12 and IL-18 and is necessary for production of IFN gamma. In T cells, IkBz is not necessary for IFN gamma and IL-4 production by Th1 and Th2 cells, respectively. However in Th17 cells, IkBz acts in concert with RORa and RORgt to upregulate IL-17A, IL-17F, IL-21, IL-22 and IL-23R expression. Applications Reported: This LK2NAP antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This LK2NAP antibody has been tested by intracellular staining and flow cytometric analysis of LPS-activated mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP...
I-kappa-B-zeta (also known as MAIL and INAP) is an ankyrin-repeat-containing nuclear protein that is highly homologous to the IkappaB family member Bcl-3. Transcription of IkappaBzeta is upregulated by stimulation with TLR ligands, IL-1 and IL-6 in cultured B-lymphocytes and monocytes/macrophages, but only faintly so in T-lymphocytes, fibroblasts, and endothelial cells. IkappaB-zeta preferentially associates with the NF-kappaB subunit p50 rather than p65 and recombinant IkappaB-zeta proteins inhibit the DNA binding of the p65/p50 heterodimer and the p50/p50 homodimer.Specifications
IkB zeta | |
0.2 mg/mL | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9EST8 | |
80859 | |
Primary | |
Affinity chromatography |
LK2NAP | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Mouse | |
NFKBIZ | |
AA408868; FLJ30225; FLJ34463; Ikappa B-zeta variant 3; ikappaBzeta; I-kappa-B-zeta; IkappaB-zeta; IKBZ; ikB-zeta; IL-1 inducible nuclear ankyrin-repeat protein; INAP; Mail; molecule possessing ankyrin repeats induced by lipopolysaccharide; molecule possessing ankyrin-repeats induced by lipopolysaccharide; NF-kappa-B inhibitor zeta; NFKB inhibitor zeta; NFKBIZ; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta; OTTHUMP00000214340; RGD1310834 | |
NFKBIZ | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.