Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKBIP Polyclonal Antibody
IKBIP Polyclonal Antibody
Supplier: Thermo Scientific PA550430
Description
IKBIP Polyclonal Antibody for IHC (P)

Specifications
IKBIP | |
Polyclonal | |
Unconjugated | |
IKBIP | |
1200009F10Rik; 1700023M03Rik; D18354; I kappa B kinase interacting protein; i kappa-B kinase interacting protein; i kappa-B kinase-interacting protein; Ikbip; IKBKB interacting protein; IKBKB-interacting protein; Ikip; IKK interacting protein; IKK-interacting protein; inhibitor of nuclear factor kappa-B kinase-interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
121457 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q70UQ0 | |
IKBIP | |
Fusion protein corresponding to a region derived from internal residues of human IKBKB interacting protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction