Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IKBIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565219 Supplier Invitrogen™ Supplier No. PA565219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSTLRNIRTVK RQEEEDLLRV EEQLGSDTKA IEKLEEEQHA LFARDEDLTN KLSDYEPKVE ECKTHLPTIE SAI Highest antigen sequence identity to the following orthologs - mouse 85%, rat 84%.

IKIP (Inhibitor of nuclear factor kappa-B kinase-interacting protein, IKK-interacting protein) is a single-pass membrane protein that shares a common promoter with APAF1. APAF1 and IKIP are both induced by X irradiation, however, the two gene products are transcribed in different directions. The IKIP gene is believed to be a target for p53 as expression of IKIP has been shown to promote apoptosis. IKIP has four known isoforms, three of which are found traversing the endoplasmic reticulum membrane. IKIP isoform 4 has a deletion of the transmembrane region which leads to a homogenous distribution of the protein within the cell. The IKIP gene products are expressed in vascular endothelial cells, while the isoform 4 has also been detected in lung, kidney, spleen, thymus and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKBIP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IKBIP
Gene Accession No. Q70UQ0
Gene Alias 1200009F10Rik; 1700023M03Rik; D18354; I kappa B kinase interacting protein; i kappa-B kinase interacting protein; i kappa-B kinase-interacting protein; Ikbip; IKBKB interacting protein; IKBKB-interacting protein; Ikip; IKK interacting protein; IKK-interacting protein; inhibitor of nuclear factor kappa-B kinase-interacting protein
Gene Symbols IKBIP
Host Species Rabbit
Immunogen Recombinant Human IKBIP. Recombinant protein control fragment (Product #RP-104692).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121457
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.