Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKBKAP Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171918AF647
Description
IKBKAP Polyclonal antibody specifically detects IKBKAP in Human samples. It is validated for Western BlotSpecifications
IKBKAP | |
Polyclonal | |
Western Blot | |
DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1 | |
The immunogen for this product maps to a region between residue 1150 and 1200 of human Inhibitor of Kappa Light Polypeptide Gene Enhancer in B-cells, Kiinase Complex-Associated Protein using the numbering given in entry NP_003631.2 (GeneID 8518). | |
0.1 mL | |
Cancer, Transcription Factors and Regulators | |
8518 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction