Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKBKAP Antibody - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$182.00 - $524.00
Specifications
Antigen | ACE/CD143 |
---|---|
Clone | MM0072-12J16 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
ACE/CD143 Monoclonal specifically detects ACE/CD143 in Human samples. It is validated for Western Blot.Specifications
ACE/CD143 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P12821.1 | |
1636 | |
IgG1 | |
Protein G purified |
MM0072-12J16 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
ACE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title