Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-9 Antibody (MM0292-4D25) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | FGF-9 |
---|---|
Clone | MM0292-4D25 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
FGF-9 Monoclonal specifically detects FGF-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FGF-9 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P31371 | |
2254 | |
IgG1 | |
Protein G purified |
MM0292-4D25 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3 | |
FGF9 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title