Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKBKAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | IKBKAP |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
IKBKAP Polyclonal antibody specifically detects IKBKAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
IKBKAP | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Transcription Factors and Regulators | |
PBS (pH 7.2), 40% Glycerol | |
8518 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RLHVLCQGWHYLAYDWHWTTDRSVGDNSSDLSNVAVIDGNRVLVTVFRQTVVPPPMCTYQLLFPHPVNQVTFLAHPQKSNDLAVLDASNQISVYKCGDCPSADPTVKLGAVG | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title