Learn More
IKBKE Mouse anti-Human, Clone: OTI1E4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807631

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IKBKE is a noncanonical I-kappa-B kinase (IKK) that is essential for regulating antiviral signaling pathways. IKBKE has also been identified as a breast cancer oncogene and is amplified and overexpressed in over 30% of breast carcinomas and breast cancer cell lines.Specifications
IKBKE | |
Monoclonal | |
Unconjugated | |
IKBKE | |
IKK-E, IKK-i, IKKE, IKKI | |
Mouse | |
Affinity Chromatography | |
RUO | |
9641 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14164 | |
IKBKE | |
Human recombit protein fragment corresponding to amino acids 474-716 of human IKBKE produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.