Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKBKE Mouse anti-Human, Clone: OTI3C5, lyophilized, TrueMAB™
SDP

Catalog No. 501668381
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8381 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8381 Supplier OriGene Supplier No. CF807632
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

IKBKE is a noncanonical I-kappa-B kinase (IKK) that is essential for regulating antiviral signaling pathways. IKBKE has also been identified as a breast cancer oncogene and is amplified and overexpressed in over 30% of breast carcinomas and breast cancer cell lines.

Specifications

Antigen IKBKE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C5
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene IKBKE
Gene Accession No. Q14164
Gene Alias IKK-E, IKK-i, IKKE, IKKI
Gene Symbols IKBKE
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 474-716 of human IKBKE produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9641
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.