Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKEPP Antibody, Novus Biologicals™
SDP

Catalog No. p-5066337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400150SS 0.025 mL
NB400150 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400150SS Supplier Novus Biologicals Supplier No. NB400150SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

IKEPP Polyclonal specifically detects IKEPP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative).

Specifications

Antigen IKEPP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry reported in scientific literature (PMID 24391715), Immunohistochemistry-Paraffin, ICC/IF (Negative)
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. Q99MJ6
Gene Alias FLJ22756, IKEPPPDZK2, intestinal and kidney enriched PDZ protein, Intestinal and kidney-enriched PDZ protein, Na(+)/H(+) exchange regulatory cofactor NHE-RF4, Na/Pi cotransporter C-terminal-associated protein 2, naPi-Cap2, Natrium-phosphate cotransporter IIa C-terminal-associated protein 2, NHERF4, NHERF-4, PDZ domain containing 2, PDZ domain containing 3, PDZ domain-containing protein 2, PDZ domain-containing protein 3, Sodium-hydrogen exchanger regulatory factor 4
Gene Symbols PDZD3
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of mouse IKEPP. [UniProt# Q99MJ6]
Molecular Weight of Antigen 60 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79849
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.