Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKEPP Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | IKEPP |
---|---|
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
IKEPP Polyclonal antibody specifically detects IKEPP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
IKEPP | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
79849 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
FLJ22756, IKEPPPDZK2, intestinal and kidney enriched PDZ protein, Intestinal and kidney-enriched PDZ protein, Na(+)/H(+) exchange regulatory cofactor NHE-RF4, Na/Pi cotransporter C-terminal-associated protein 2, naPi-Cap2, Natrium-phosphate cotransporter IIa C-terminal-associated protein 2, NHERF4, NHERF-4, PDZ domain containing 2, PDZ domain containing 3, PDZ domain-containing protein 2, PDZ domain-containing protein 3, Sodium-hydrogen exchanger regulatory factor 4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: PSTLSGHRVCQAHGEPVLGLCPLLPLFCCPPHPPDPWSLERPRFCLLSKEEGKSFGFHLQQELGRAGHVVCRVDPGTSAQRQGLQEGDRILAVNNDV | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title