Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ IKK 16
GREENER_CHOICESDP

Catalog No. 253950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-395-0 50 mg
25-391-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-395-0 Supplier R&D Systems™ Supplier No. 2539/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of IKK

Selective inhibitor of IκB kinase (IKK) (IC50 values are 40, 70 and 200 nM for IKKβ, IKK complex and IKKα respectively). Inhibits TNF-α-stimulated IκB degradation and expression of adhesion molecules E-selectin, ICAM and VCAM (IC50 values are 1.0, 0.5, 0.3 and 0.3 μM respectively). Inhibits LPS-induced TNF-α release in vivo and neutrophil extravasion in thioglycollate-induced peritonitis.

Chemical Identifiers

CAS 1186195-62-9
Molecular Formula C28H30ClN5OS

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 520.09
Purity >98%
Molecular Weight (g/mol) 520.092
Chemical Name or Material IKK 16
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.