Learn More
Promega IKK alpha Kinase Enzyme System, 10microg
Catalog No. PRV4068 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4068 10 μg
1 options
Catalog No. PRV4068 Supplier Promega Supplier No. V4068
Full-length recombinant human IKK alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. IKK alpha is a serine/threonine protein kinase that phosphorylates the I-kappa-B protein.
  • Easily Screen and Profile IKKα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C