Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IKK alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585942 Supplier Invitrogen™ Supplier No. PA585942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IKK alpha Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

CHUK (IKK alpha) is a serine/threonine kinase involved in signal transduction and cellular communication. It belongs to the cytokine activated protein complex that is an inhibitor of the NF-kappa B complex. IKK alpha along with IKK beta and a regulatory protein NEMO (IKK gamma) phosphorylates IkB proteins which targets the later for ubiquitin mediated degradation. It is involved with several cellular processes with extensive studies on its role in inflammation and cell survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKK alpha
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene CHUK
Gene Accession No. O15111, Q60680
Gene Alias AI256658; CHUK; Chuk1; component of inhibitor of nuclear factor kappa B kinase complex; conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; Fbx24; Fbxo24; FLJ40509; I kappa B kinase 1; i kappa-B kinase alpha; ikappaB kinase; I-kappa-B kinase 1; I-kappa-B kinase 2; IkappaB kinase alpha; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; IkB kinase alpha subunit; IkB kinase-alpha; ikBKA; IKBKB; IKK alpha; IKK1; IKK2; Ikka; IKK-A; IKK-a kinase; IKK-alpha; IKKB; IKK-B; IKK-beta; Inhibitor of nuclear factor kappa-B kinase subunit alpha; kinase beta; MGC131801; NFKBIKA; NFKBIKB; Nuclear factor NFkappaB inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase alpha; TCF16; TCF-16; Transcription factor 16
Gene Symbols CHUK
Host Species Rabbit
Immunogen Synthetic peptide within Human Ikk alpha aa 1-50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1147, 12675, 309361
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.