Learn More
Invitrogen™ IKK alpha Polyclonal Antibody

Description
Human IKK alpha shares 98% amino acid (aa) sequence identity with mouse IKK alpha. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human K562 whole cell, human HepG2 whole cell, human Caco-2 whole cell. IHC: human lung cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | IKK alpha |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5mg BSA and 0.05mg sodium azide |
| Gene | CHUK |
| Gene Accession No. | O15111 |
| Gene Alias | AI256658; CHUK; Chuk1; component of inhibitor of nuclear factor kappa B kinase complex; conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; Fbx24; Fbxo24; FLJ40509; I kappa B kinase 1; i kappa-B kinase alpha; ikappaB kinase; I-kappa-B kinase 1; I-kappa-B kinase 2; IkappaB kinase alpha; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; IkB kinase alpha subunit; IkB kinase-alpha; ikBKA; IKBKB; IKK alpha; IKK1; IKK2; Ikka; IKK-A; IKK-a kinase; IKK-alpha; IKKB; IKK-B; IKK-beta; Inhibitor of nuclear factor kappa-B kinase subunit alpha; kinase beta; MGC131801; NFKBIKA; NFKBIKB; Nuclear factor NFkappaB inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase alpha; TCF16; TCF-16; Transcription factor 16 |
| Gene Symbols | CHUK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.