Learn More
IKK-alpha Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301934AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
Inhibitor of Nuclear factor Kappa-B kinase subunit alpha (IKK alpha) is encoded by the CHUK gene and located on chromosome 10 in humans. It is also known as IKK1 or CHUK (Conserved helix-loop-helix ubiquitous kinase). IKK alpha belongs to the family of serine threonine protein kinase and forms a part of IKK complex. The IKK complex is required for the activation of NF-kB with IKK alpha specifically known to be involved in the non-canocial pathway for NF-kB activation. IKK alpha along with IKK beta and a regulatory protein NEMO (IKK gamma) phosphorylates IkB proteins which targets the later for ubiquitin mediated degradation. It is involved with several cellular processes with extensive studies on its role in inflammation and cell survival.Specifications
IKK-alpha | |
Polyclonal | |
Unconjugated | |
CHUK | |
IKBKA, IKK-alpha, IKK1, IKKA, NFKBIKA, TCF16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1147 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O15111 | |
CHUK | |
Between 550 and 600 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.