Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK beta Antibody (10AG2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280788
Description
IKK beta Monoclonal specifically detects IKK beta in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, CyTOF-ready.Specifications
IKK beta | |
Monoclonal | |
1 mg/mL | |
Western Blot : 2 - 4 ug/ml, Simple Western : 1:12.5, Flow Cytometry : 0.25 - 0.5 ug/ 10^6 cells, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2-5 ug/ml. Use reported in scientific literature (PMID 24825920), Immunoprecipitation : 1:10 - 1:500, Immunohistochemistry-Paraffin reported in scientific literature (Page et al), Immunoblotting reported in scientific literature (PMID 20103608), CyTOF-ready | |
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
10AG2 | |
Unconjugated | |
PBS | |
IKBKB | |
A full-length human IKKb recombinant protein was used to immunize mice (NP_001547). | |
0.1 mg | |
Cancer, Immunology, Innate Immunity, mTOR Pathway, Phospho Specific, Protein Kinase | |
3551 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction