Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK beta Antibody (10AG2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056509R
Description
IKK beta Monoclonal specifically detects IKK beta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IKK beta | |
Monoclonal | |
DyLight 550 | |
O14920 | |
IKBKB | |
A full-length human IKKb recombinant protein was used to immunize mice (NP_001547). | |
Protein G purified | |
RUO | |
Primary | |
An approx. 87 kDa band should be observed. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
10AG2 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta | |
Mouse | |
86.564 kDa | |
0.1 mL | |
Cancer, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase | |
3551 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction