Learn More
Promega IKK beta Kinase Enzyme System, 1mg
Catalog No. PRV6587 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6587 1 mg
1 options
Catalog No. PRV6587 Supplier Promega Supplier No. V6587
Add to Cart
Add to Cart
Request a Quote
Full length recombinant human IKKβ was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile IKKβ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C