Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    IKK epsilon/IKBKE Antibody (OTI3C5), Allophycocyanin, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271490APC
Description
IKK epsilon/IKBKE Monoclonal antibody specifically detects IKK epsilon/IKBKE in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| IKK epsilon/IKBKE | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Western Blot, Immunohistochemistry | |
| OTI3C5 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase epsilon, IkBKE, IKK-E, IKKEMGC125297, IKK-epsilon, IKKI, IKK-iMGC125295, Inducible I kappa-B kinase, inducible IkappaB kinase, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon, inhibitor of nuclear factor kappa-B kinase subunit epsilon, KIAA0151IKK-related kinase epsilon, MGC125294 | |
| Human recombinant protein fragment corresponding to amino acids 474-716 of human IKBKE(NP_054721) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Phospho Specific, Protein Kinase | |
| 9641 | |
| Store at 4C in the dark. | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction