Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK epsilon/IKBKE Antibody (OTI3C5), Janelia Fluor™ 549, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271490JF549

Catalog No. N271490J549


Only null left
Add to Cart

Description

Description

IKK epsilon/IKBKE Monoclonal specifically detects IKK epsilon/IKBKE in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

IKK epsilon/IKBKE
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
IKBKE
Recombinant protein fragment corresponding to amino acids 474-716 of human IKK epsilon/IKBKE (NP_054721) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI3C5
Western Blot, Immunohistochemistry
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase epsilon, IkBKE, IKK-E, IKKEMGC125297, IKK-epsilon, IKKI, IKK-iMGC125295, Inducible I kappa-B kinase, inducible IkappaB kinase, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon, inhibitor of nuclear factor kappa-B kinase subunit epsilon, KIAA0151IKK-related kinase epsilon, MGC125294
Mouse
Affinity Purified
Cancer, Phospho Specific, Protein Kinase
9641.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.