Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK epsilon/IKBKE, p Thr501 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177829JF646
Description
IKK epsilon/IKBKE Polyclonal specifically detects IKK epsilon/IKBKE in Human samples. It is validated for ELISA.Specifications
IKK epsilon/IKBKE | |
Polyclonal | |
ELISA | |
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase epsilon, IkBKE, IKK-E, IKKEMGC125297, IKK-epsilon, IKKI, IKK-iMGC125295, Inducible I kappa-B kinase, inducible IkappaB kinase, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon, inhibitor of nuclear factor kappa-B kinase subunit epsilon, KIAA0151IKK-related kinase epsilon, MGC125294 | |
IKK epsilon/IKBKE phospho peptide corresponding to a region of the human protein surrounding pT501 conjugated to KLH. (Uniprot: Q14164) | |
0.1 mL | |
Cancer, Phospho Specific, Protein Kinase | |
9641 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction