Learn More
Invitrogen™ IKK epsilon Recombinant Rabbit Monoclonal Antibody (5N4J8)

Description
Immunogen sequence: MQSTANYLWH TDDLLGQGAT ASVYKARNKK SGELVAVKVF NTTSYLRPRE VQVREFEVLR KLNHQNIVKL FAVEETGGSR QKVLVMEYCS SGSLLSVLES.
Specifications
Specifications
| Antigen | IKK epsilon |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 5N4J8 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | IKBKE |
| Gene Accession No. | Q14164, Q9R0T8 |
| Gene Alias | AW558201; I-kappa-B kinase epsilon; Ikbke; IKKE; IKK-E; IKKepsilon; IKK-epsilon; IKKI; IKK-i; IKK-related kinase epsilon; inducible I kappa-B kinase; inducible IkappaB kinase; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon; inhibitor of kappaB kinase epsilon; inhibitor of nuclear factor kappa-B kinase subunit epsilon; KIAA0151; RP11-534L20.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.