Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK gamma Antibody (46B844), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056532AF750
Description
IKK gamma Monoclonal specifically detects IKK gamma in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
IKK gamma | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
IKBKG | |
Full-length human IKK gamma protein. | |
0.1 mL | |
Primary | |
A 48 kDa band is observed. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
46B844 | |
Western Blot, Flow Cytometry | |
FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator | |
Mouse | |
Protein G purified | |
Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific | |
8517.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction